Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 1,450, waxy soft solid, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

BAKRU220-07 BAKRU220-07
JTU220-7CS 2381.1 USD InStock
CAJTU220-7 JTU220-7
Polyethylene glycol 1,450, waxy soft solid, J.T.Baker®
Polyethylene glycol 1,450


Formula: H(OCH₂CH₂)nOH
MW: 1450 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty