Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 8,000, crystalline powder, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: PEG 8,000

Management of Change (MOC) category = N

BAKRU222-08 BAKRU222-08 BAKRU222-09 BAKRU222-01 BAKRU222-09 BAKRU222-01
JTU222-8CS 1067.98 USD InStock
JTU222-8 JTU222-9 CAJTU222-1 JTU222-1 CAJTU222-9 CAJTU222-8
Polyethylene glycol 8,000, crystalline powder, J.T.Baker®
Polyethylene glycol 8,000
Powder.


Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250 °C (1013 hPa)
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Viscosity, cSt 470 - 900
Average Molecular Weight 7000 - 9000

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty