Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionic acid ≥99% (by GC), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Propanoic acid

Management of Change (MOC) category = N

BAKRU330-09 BAKRU330-07
JTU330-9CS 911.81 USD InStock
JTU330-9 JTU330-7
Propionic acid ≥99% (by GC), J.T.Baker®
Propionic acid


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: –21 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8 (3),II
REACH: 01-2119486971-24
Merck Index: 12,08010
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (CH₃CH₂COOH) (by GC) ≥99 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty