Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropyl acetate ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Acetic acid isopropyl ester

Management of Change (MOC) category = N

BAKRU385-26 U385-07 BAKRU385-07 BAKRU385-6K BAKRU385-03 BAKRU385-6K BAKRU385-03 BAKRU385-26
JTU385-7EA 1910.46 USD InStock
JTU385-7 CAJTU385-3 CAJTU385-26 JTU385-6K JTU385-3 CAJTU385-7 CAJTU385-6K JTU385-26
Isopropyl acetate ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Isopropyl acetate
Lot analysis on label.


Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 85 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.8732 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008877
CAS Number: 108-21-4
EINECS: 203-561-1
UN: 1220
ADR: 3,II
Merck Index: 13,05225
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (by GC) ≥99.0 %
Density (g/mL) at 25°C 0.86 - 0.87
Residue after Evaporation ≤0.0050 %
Acidity (as CH₃COOH) ≤0.01 %
Water (H₂O) ≤0.1 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty