Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Schiff reagent (Hotchkiss & McManus), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = NI

BAKRU973-01 BAKRU973-01
JTU973-1CS 2653.02 USD InStock
JTU973-1 CAJTU973-1
Schiff reagent (Hotchkiss & McManus), J.T.Baker®
Schiff reagent
For Periodic Acid-Schiff Reaction.

Contains 4.54 g/L basic fuchsin, 4.543 g/L sodium bisulphite, and 7.5 ml/L hydrochloric acid in water.


Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
pH at 25°C 1.1 - 1.5
Absorbance at 545 nm (10-mm path vs water) ≤0.020
Sensitivity for Aldehyde Passes Test
Suitability for Periodic Acid-Schiff Staining Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty