Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKRV398-08 BAKRV398-08
JTV398-8EA 385.47 USD InStock
CAJTV398-8 JTV398-8
1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®
1,1,2,2-Tetrachloroethane


Formula: Cl₂HCCHCl₂
MW: 167.85 g/mol
Boiling Pt: 146.3 °C (1013 hPa)
Melting Pt: –43 °C
Density: 1.595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000848
CAS Number: 79-34-5
EINECS: 201-197-8
UN: 1702
ADR: 6.1,II
Merck Index: 12,09331
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/34/725334.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (CHCl₂CHCl₂) (by GC) ≥98 %
Appearance Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty