Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Trizma(R) baseTRIS baseTHAM2-Amino-2-(hydroxymethyl)-1,3-propanediolTris(hydroxymethyl)methylamine2-Amino-2-(hydroxymethyl)propane-1,3-diol2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris)Tromethamine

Management of Change (MOC) category = N

BAKRX171-07 X171-03 BAKRX171-05 BAKRX171-05 BAKRX171-07 BAKRX171-03
JTX171-3EA 715.81 USD InStock
CAJTX171-3 CAJTX171-5 CAJTX171-7 JTX171-3 JTX171-5 JTX171-7
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Lot analysis on label.


Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 12,09902
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/00/2297600.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₄H₁₁NO₃) (dried basis) 99.8 - 100.1 %
Absorbance of a 40% Solution: Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤2 %
Insoluble Matter ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
pH of 0.1M Solution at 25°C, For Information Only
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty