Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Viral inactivation solution, Ultrapure Bioreagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


BAKRX191-04 BAKRX191-04 BAKRX191-01 BAKRX191-01 BAKRX191-09
JTX191-04EA 220.69 USD InStock
JTX191-04 CAJTX191-01 JTX191-01 JTX191-09 CAJTX191-04
Viral inactivation solution, Ultrapure Bioreagent, J.T.Baker®
Viral inactivation solution
J.T.Baker® Viral Inactivation Solution Ultrapure bioreagent is a highly effective, ready-to-use product, specifically developed for detergent-based viral inactivation in biopharmaceutical operations and plasma derived therapy processes. J.T.Baker® Viral Inactivation Solution is formulated with non-ionic detergent, that is readily biodegradable and alkylphenol ethoxylate free, addressing environmental concerns associated with REACH. This innovative solution is optimized to have low protein reactive impurities, low viscosity and compatibility with typical downstream processes. J.T.Baker® Viral Inactivation Solution Ultrapure bioreagent is available in a variety of sizes to support development to large scale commercial batches. Detail description provided in technical literature.

  • High purity, tested for bioburden and impurity
  • Effective inactivation of virus
  • Easily cleared in downstream processing, detectable to 1ppm
  • Readily biodegradable
  • 0.2 µm filtered solution

Proprietary Formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)


Order Now
Specification Test Results
Identification (by FTIR) Passes Test
Appearance Passes Test
Purity by HPLC min 95%
Assay Passes Test
Acid value max 10
Viscosity (25°C) For information only
Density 0.9 – 1.1 g/mL
Limit of free ethylene oxide (Target <1 ppm) For information only
Total microbial count (No growth detected) Passes Test
Endotoxin For information only
Protein reactive impurities (100x Diluted) For information only
Arsenic (As) For information only
Cadmium (Cd) For information only
Lead (Pb) For information only
Mercury (Hg) For information only
Cobalt (Co) For information only
Nickel (Ni) For information only
Vanadium (V) For information only
Silver (Ag) For information only
Gold (Au) For information only
Iridium (Ir) For information only
Osmium (Os) For information only
Palladium (Pd) For information only
Platinum (Pt) For information only
Rhodium (Rh) For information only
Ruthenium (Ru) For information only
Selenium (Se) For information only
Thallium (Tl) For information only
Barium (Ba) For information only
Chromium (Cr) For information only
Copper (Cu) For information only
Lithium (Li) For information only
Molybdenum (Mo) For information only
Antimony (Sb) For information only
Tin (Sn) For information only

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty