Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polysorbate 80, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Polyoxyethylene-80-Sorbitan MonolauratePolyoxyethylene (20) sorbitan monooleateSorbitan monooleate, ethoxylated

Management of Change (MOC) category = N

BAKRX257-07 BAKRX257-09 X257-09 BAKRX257-01 BAKRX257-07 BAKRX257-01
JTX257-9CS 10568.05 USD InStock
CAJTX257-7 JTX257-9 JTX257-7 CAJTX257-1 JTX257-01 CAJTX257-9
Polysorbate 80, J.T.Baker®
Polysorbate 80
Polyoxyethylene (20) Sorbitan Monooleate


Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-018-3
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test
Not intended for Injectable Drug Use.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty