Would you like to log in again?
KIMBLE® Glass Wash Bottle, 1000 ml, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
1011
KT1011EA 260.6 USD InStock
KT1011
KIMBLE® Glass Wash Bottle, 1000 ml, DWK Life Sciences
Bottles Wash Bottles
Unitized Wash bottles allow you to automatically pump a continuous, forceful jet of washing or rinsing liquid that may be instantly stopped as needed.
  • Easy one-handed operation (either hand), leaving the other hand free for other needs
  • Crystal clear glass flask permits quick inspection of the contents
  • Unitized Wash bottles are recommended for use at ambient temperatures
  • Features borosilicate glass flask and neoprene parts for improved solvent resistance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty