Would you like to log in again?
Vacuum Filtration Adapter with PTFE Stopcock, DWK Life Sciences
Supplier: DWK Life Sciences
178130-2942
KT178130-2942EA 257.4 USD InStock
KT178130-2942
Vacuum Filtration Adapter with PTFE Stopcock, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Vacuum filtration adapters with PTFE stopcock is used for reduced pressure filtration using plain stem buchner funnels.
  • PTFE stopcock permits control of vacuum source
  • Designed with a flange to accommodate a pluro stopper
  • Replacement PTFE stopcock plug is 821001 series
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Pluro stopper sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty