Would you like to log in again?
KIMBLE® BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences
BEVEL-SEAL™
179700-1424 746030-0510 746030-0000 179700-1729 179700-0519 179700-2119 179700-2129 179700-1124 179700-0524 179700-0514 179700-2124 179700-0819 179700-2114 179700-0814 179700-0824 179700-0529
10000-500EA 117.33 USD InStock
10000-500 KT746030-0000 KT179700-0819 KT746030-0510 KT179700-0529 KT179700-0519 KT179700-0514 KT179700-0824 KT179700-0814 KT179700-1424 KT179700-2119 KT179700-2129 KT179700-0524 KT179700-2124 KT179700-1124 KT179700-2114
KIMBLE® BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
BEVEL-SEAL™ Inlet Adapters are designed for use with plain thermometers, pipets, syringes, or other small items with an O.D. from 2 to 17mm.
  • Vacuum-tight seal
  • Open-top compression cap and FKM O-Ring included
  • Accommodates adjustable immersion of thermometers
A vacuum-tight seal is created with the supplied open-top compression cap and FKM O-Ring to allow adjustable immersion of thermometers. Ref: ASTM Method D1744.
The cap is suitable for use to 150 °C (302 °F). To use it as a septum port, replace the O-Ring with a PTFE-lined septum.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty