Would you like to log in again?
KIMBLE® BEVEL-SEAL™ Thermometer Inlet Adapters with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences
BEVEL-SEAL™
179750-2129 746035-0814 179750-0829 746035-0510 179750-0824 179750-2124 179750-2114 179750-0524 179750-1119 179750-1129 179750-0814 179750-0514
KT179750-1129CS 141.2 USD InStock
KT179750-1129 KT179750-1119 KT179750-0824 KT179750-0514 KT179750-2129 KT746035-0814 KT179750-0524 KT179750-2124 KT179750-2114 KT179750-0814 KT746035-0510 KT179750-0829
KIMBLE® BEVEL-SEAL™ Thermometer Inlet Adapters with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed for use with plain thermometers, pipets, syringes, or other small items with an O.D. from 2mm to 11mm.
  • Vacuum-tight seal
  • With hose connection
  • One FKM O-Ring and one cap included
Equipped with a hose connection for vacuum or the introduction of gases
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
A vacuum-tight seal is created with the open-top compression cap and FKM O-Ring to allow adjustable immersion of thermometers, etc. Cap is suitable for use to 150°C (302°F). To use as a septum port, replace the O-Ring with a PTFE-lined septum. Supplied with one FKM O-Ring and one cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty