Would you like to log in again?
KIMBLE® PTFE BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences
BEVEL-SEAL™
179800-2214 179800-2324 179800-2124 179800-2224
KT179800-2214EA 167.9 USD InStock
KT179800-2214 KT179800-2324 KT179800-2124 KT179800-2224
KIMBLE® PTFE BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
This is an adjustable fitting designed to adapt to a variety of apparatus.
  • Vacuum-tight seal
  • Bottom [ST] inner joint
  • Design allows use with clamps
  • FKM O-Rings and polypropylene cap included
A diameter variation of 1mm to 2mm can be accommodated, as the inert O-Ring is compressed by the open-top cap to create a vacuum-tight seal. The adapter body is made of PTFE. Bottom has a Standard Taper inner joint. Design allows use with clamps. Supplied with FKM O-Rings and with polypropylene cap suitable for use to 150°C (302°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty