Would you like to log in again?
KIMBLE® KONTES® Universal Inlet with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences
KONTES®
179850-2214 179850-2124 179850-2114 179850-2224 179850-2324
KT179850-2124EA 195.23 USD InStock
KT179850-2124 KT179850-2214 KT179850-2324 KT179850-2114 KT179850-2224
KIMBLE® KONTES® Universal Inlet with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed to adapt a variety of tubes, condensers and filters in 14/20 and 24/25–24/40 glassware.
  • Inert O-ring
  • Vacuum tight seal
  • Serrated side port
  • Bottom inner joint
Diameter variations of 1 to 2mm can be accommodated, as the inert O-ring is compressed by the open-top polypropylene cap to effect a vacuum tight seal. The serrated side port can be used for a vacuum connection or for the introduction of gases. Bottom has an inner joint. Body is PTFE, O-rings are Viton, polypropylene compression cap is suitable for use to 150ºC, and side port is CTFE.
Ordering information: Includes Compression Cap.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty