Would you like to log in again?
KIMBLE® AIRLESS-WARE® Manifold, DWK Life Sciences
Supplier: DWK Life Sciences
KONTES® AIRELSS-WARE®
216060-0003 216060-0000
KT216060-0000EA 1892.15 USD InStock
KT216060-0000 KT216060-0003
KIMBLE® AIRLESS-WARE® Manifold, DWK Life Sciences
Manifolds
Used to connect inert gas and vacuum sources
The 4 mm (0.157") PTFE vacuum valves provide grease-free fine control. Sample contacts only PTFE and borosilicate glass. Distance between valves is 100 mm (3.94"). Beams have a 12.7 mm (0.5") outer diameter (O.D.), delivery ports have a 9 mm (0.35") O.D.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty