Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Vacuum Manifolds, Inert Gas, Double, with Vacuum Chambers, DWK Life Sciences

Supplier: DWK Life Sciences
KIMBLE®
216130-0003 216130-0005 216130-0004
KT216130-0005EA 2272.86 USD InStock
KT216130-0005 KT216130-0003 KT216130-0004
Vacuum Manifolds, Inert Gas, Double, with Vacuum Chambers, DWK Life Sciences
Manifolds
General purpose double bank manifold for use with vacuum and inert gas.

  • Medium wall tubing size: ¹/₂"
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Stopcocks are 4 mm bore glass with lower vacuum chamber and spaced 85 mm

Top right connection is for inert gas and lower left connection is for vacuum.

Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty