Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Vacuum Manifolds, Inert Gas, with PTFE Valves, DWK Life Sciences

Supplier: DWK Life Sciences
KIMBLE®
216140-0003 216140-0004 216140-0005
KT216140-0003EA 1372.75 USD InStock
KT216140-0003 KT216140-0005 KT216140-0004
Vacuum Manifolds, Inert Gas, with PTFE Valves, DWK Life Sciences
Manifolds
These manifolds are used to connect inert gas and vacuum sources to sidearm stopcocks (three-place, four-place or five-place manifolds). Which is manufactured from 33 expansion, low extractable borosilicate glass.

  • Distance between valves is 100 mm
  • Ports have 9 mm OD openings
  • Sample contacts only PTFE and borosilicate glass

Each stopcock has a metering groove cut into the plug to provide fine control of vacuum sources and inert gases. Beams are ¹/₂" (12.7 mm) OD, with upper-left and lower-right ends open.

Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty