Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Vacuum Manifolds, Inert Gas, Double, with PTFE Valves, DWK Life Sciences

Supplier: DWK Life Sciences
KIMBLE®
216145-0004 216145-0005
KT216145-0004EA 1976.76 USD InStock
KT216145-0004 KT216145-0005
Vacuum Manifolds, Inert Gas, Double, with PTFE Valves, DWK Life Sciences
Manifolds
Double vacuum gas manifold is for connecting inert gas and vacuum sources.

  • Medium wall tubing: ³/₄"
  • Rob braces gives additional strength
  • Opened upper left and lower right beams
  • Manufactured from 33 expansion, low extractable borosilicate glass

100 mm spaced ports with serrated hose connections.

PTFE vacuum valves (4 mm) at 45° angle is for grease free operation and to allow easier manipulation.

Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty