Would you like to log in again?
KIMBLE® Inner Joint Hose Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences
182900-2440 276100-1420 276100-1922
KT276100-1922EA 63.95 USD InStock
KT276100-1922 KT276100-1420 KT182900-2440
KIMBLE® Inner Joint Hose Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
This inlet adapter is ideal for use as a vent, or for the introduction of pressure/vacuum.
  • Inner [ST] joint at one end
  • Vertical hose connection at other end
  • Manufactured from borosilicate glass
Adapter features an inner Standard Taper joint at one end and vertical hose connection at the other.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty