Would you like to log in again?
KONTES® Reflux Condenser, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KONTES®
283000-0000
KT283000-0000EA 412.79 USD InStock
KT283000-0000
KONTES® Reflux Condenser, Kimble Chase
Condensers
With an internal, coil-type cold finger
Approximate overall height: 200mm (77/8").
[ST] Outer joints at the top, [ST] Inner drip joint at the bottom and 2 hose connectors at the top for water inlet and outlet. Tightly wound coil provides enough surface area to condense high vapor pressure solvents like hexane. #1 hose connections.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty