Would you like to log in again?
KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
292010-0000 292011-0000
KT292010-0000EA 604.6 USD InStock
KT292010-0000 KT292011-0000
KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
Extractors Soxhlet and Soxhlet-Type Extractors
Soxhlet-type extraction apparatus is for use with extraction thimble, which has a coarse porosity.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty