Would you like to log in again?
KIMBLE® Distilling Flasks, Round Bottom, 3-Neck, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
295550-0250 295510-0100 295510-0500 295500-0250 295550-0100 295510-0050 295500-0100 295500-0050 295550-0500 295550-0050 295550-0015 295510-0250 295500-0015 295550-0025
KT295550-0250EA 169.91 USD InStock
KT295550-0250 KT295500-0100 KT295550-0050 KT295510-0500 KT295500-0250 KT295500-0050 KT295550-0500 KT295510-0100 KT295550-0015 KT295550-0025 KT295550-0100 KT295510-0250 KT295500-0015 KT295510-0050
KIMBLE® Distilling Flasks, Round Bottom, 3-Neck, DWK Life Sciences
Flasks Distillation Flasks
Angled three neck round bottom flasks with standard taper outer joints.
  • Side necks are angled
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty