Would you like to log in again?
Separating Funnels, DWK Life Sciences
Supplier: DWK Life Sciences
298281-0010 298281-0025 298280-0125 298280-0025 298280-0010
KT298280-0010EA 256.27 USD InStock
KT298280-0010 KT298280-0125 KT298280-0025 KT298281-0010 KT298281-0025
Separating Funnels, DWK Life Sciences
Funnels Separating Funnels
Micro separating funnels manufactured from 33 expansion, low extractable borosilicate glass
Squibb-type funnel with a PTFE stopcock plug with a 2 mm bore.
Certifications: Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Series 298280 is supplied with hex-head stopper and series 298281 is supplied without stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty