Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Precision Ground Stirrer Shaft, Glass, Kimble Chase
Supplier: Kimble Chase
786500-0263 786500-0264 786500-0262 786500-0261 304151-0000
KT786500-0262EA 126.45 USD InStock
KT786500-0262 KT786500-0263 KT786500-0261 KT304151-0000 KT786500-0264
Precision Ground Stirrer Shaft, Glass, Kimble Chase
Stirrers Stirring Shafts
Glass stirrer shaft with a precision-ground surface
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty