Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plain Reusable Centrifuge Tubes with Screw Caps, Kimble Chase

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

410090-0015 410090-0050
KT410090-0050PK 484.08 USD InStock
KT410090-0050 KT410090-0015
Plain Reusable Centrifuge Tubes with Screw Caps, Kimble Chase
Tubes Centrifuge Tubes
Threaded ungraduated conical centrifuge tube with a PTFE/rubber-lined black phenolic screw cap.

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Ordering information: The 15 ml tubes fit IEC Shield Number 302. The 50 ml tubes fit IEC Shield Number 320.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty