Would you like to log in again?
Open-Top Nylon Screw Thread Caps, Kimble Chase
Supplier: DWK Life Sciences
410119-1307 410119-3021 410119-2015 410119-3527 410119-1305 410119-2214 410119-2011 410119-1508 410119-2817 410119-3324
KT410119-1305CS 43.27 USD InStock
KT410119-1305 KT410119-1307 KT410119-3527 KT410119-2214 KT410119-2015 KT410119-3324 KT410119-2011 KT410119-3021 KT410119-1508 KT410119-2817
Open-Top Nylon Screw Thread Caps, Kimble Chase
Closures Bottle Closures
Glass-filled, blue nylon cap with modified GPI thread finish to accommodate use with an O-ring seal
Caps with 7mm and 15mm hole diameters are used for the MICROFLEX® microscale chemistry kits. The cap with 8.5mm hole diameter is a compression cap. Temperature limit is 150°C (302°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty