Would you like to log in again?
KONTES® CHROMAFLEX™ Chromatography Column with Fritted Disc, PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences
KONTES® CHROMAFLEX™
420540-0630 420540-2610 420540-0233 420540-0243 420540-0213 420540-0530 420540-0650 420540-0450 420540-0750 420540-0245 420540-0660 420540-0256 420540-0430 420540-0760 420540-0224 420540-0234
KT420540-0630EA 317.25 USD InStock
KT420540-0630 KT420540-0245 KT420540-0256 KT420540-0234 KT420540-2610 KT420540-0213 KT420540-0224 KT420540-0243 KT420540-0650 KT420540-0430 KT420540-0760 KT420540-0233 KT420540-0530 KT420540-0750 KT420540-0450 KT420540-0660
KONTES® CHROMAFLEX™ Chromatography Column with Fritted Disc, PTFE Stopcock, Kimble Chase
Chromatography Columns
Manufactured from medium-wall tubing.
Features a PTFE stopcock and a coarse porosity glass frit (40 to 60 µm) at lower end to support packing material.
Ordering information: For other sizes and plastic safety coating, contact your Avantor sales representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty