Would you like to log in again?
Chromatography Glass Gravity Columns, Kimble Chase
Supplier: DWK Life Sciences
420550-0760 420550-0325 420550-0130 420550-0234 420550-0225 420550-0320 420550-0224 420550-0245 420550-0550 420550-0660 420550-0220 420550-2610 420550-0430 420550-0750 420550-0125 420550-0630 420550-0213 420550-0230 420550-0560 420550-0730 420550-0650 420550-0425 420550-0233 420550-0450 420550-0530 420550-0330 420550-0243 420650-0560 420550-0256
KT420550-0560EA 615.1 USD InStock
KT420550-0560 KT420650-0560 KT420550-0660 KT420550-0220 KT420550-0760 KT420550-0320 KT420550-0243 KT420550-0245 KT420550-0224 KT420550-0125 KT420550-0225 KT420550-0325 KT420550-0425 KT420550-0130 KT420550-0230 KT420550-0450 KT420550-0550 KT420550-0330 KT420550-0430 KT420550-0650 KT420550-2610 KT420550-0750 KT420550-0233 KT420550-0530 KT420550-0234 KT420550-0630 KT420550-0256 KT420550-0213 KT420550-0730
Chromatography Glass Gravity Columns, Kimble Chase
Chromatography Columns
Gravity column manufactured from medium wall tubing with PTFE stopcock.
Glass frit (40 to 60 µm porosity) at the lower end to act as a support for column packing material. Custom sizes and plastic safety coating are available upon request.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty