Would you like to log in again?
Chromatography Glass Columns with Reservoir, Kimble Chase
Supplier: DWK Life Sciences
420610-0560 420610-0450 420610-0630 420610-0320 420610-0660 420610-0430 420610-0730 420610-0530 420610-0550 420610-0125
KT420610-0560EA 464.07 USD InStock
KT420610-0560 KT420610-0450 KT420610-0660 KT420610-0550 KT420610-0320 KT420610-0430 KT420610-0530 KT420610-0630 KT420610-0730 KT420610-0125
Chromatography Glass Columns with Reservoir, Kimble Chase
Chromatography Columns
Gravity chromatography column manufactured from medium wall tubing with PTFE stopcock and in-line reservoir for containing liquid buffer.
  • Standard taper joint size 24/40
Glass wool (not supplied) may be used as a support for the packing material. Custom sizes and plastic safety coating are available upon request.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty