Would you like to log in again?
KIMBLE® CHROMAFLEX® Chromatography Column Polypropylene Ferrule Fittings, DWK Life Sciences
Supplier: DWK Life Sciences
420828-0116 420828-0018
KT420828-0018CS 4.22 USD InStock
KT420828-0018 KT420828-0116
KIMBLE® CHROMAFLEX® Chromatography Column Polypropylene Ferrule Fittings, DWK Life Sciences
Fittings and Connectors
Polypropylene ferrule fitting used to make connections between the tubing and the end fittings of CHROMAFLEX® columns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty