Would you like to log in again?
CHROMAFLEX® Columns Screw Caps, Kimble Chase
Supplier: DWK Life Sciences
420833-1000 420833-0000 420833-1010 420833-0020 420833-0010 420833-1020
KT420833-0010EA 26.43 USD InStock
KT420833-0010 KT420833-0020 KT420833-1010 KT420833-1000 KT420833-0000 KT420833-1020
CHROMAFLEX® Columns Screw Caps, Kimble Chase
Closures Chromatography Vial Closures
Screw caps to be used with the CHROMAFLEX® columns
Some caps can be used with the tapered and standard columns and some caps can be used with the water-jacketed columns. For use with is designated for each article.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty