Would you like to log in again?
KONTES® CHROMAFLEX™ Safety/Water-Jacketed Chromatography Columns, Kimble Chase
Supplier: DWK Life Sciences
KONTES® CHROMAFLEX™
420870-1220 420870-1020 420870-1210 420870-6020 420870-1510 420870-6000 420870-1500 420870-1520 420870-6010 420870-3010 420870-1200 420870-3020 420870-3000 420870-1010 420870-1000
KT420870-3010EA 958.04 USD InStock
KT420870-3010 KT420870-3020 KT420870-1020 KT420870-3000 KT420870-1010 KT420870-1220 KT420870-1000 KT420870-1210 KT420870-1200 KT420870-1520 KT420870-1510 KT420870-1500 KT420870-6010 KT420870-6000 KT420870-6020
KONTES® CHROMAFLEX™ Safety/Water-Jacketed Chromatography Columns, Kimble Chase
Chromatography Columns
For medium pressure chromatography with either aqueous or organic mobile phases.
Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. Flow adapters are constructed with PTFE eluant cannula, acetal body, PTFE/propylene O-ring seal, and 20 µm polyethylene bed support.
A PTFE bed support is required when flow adapter is used with aggressive organic solvents, PTFE bed supports include 20 µm PTFE bed support, 40 mesh PTFE support screen, and PTFE retainer ring.
These threaded borosilicate glass columns feature an acrylic water jacket that provides thermal control of temperature-sensitive chromatographic separations and acts as a safety shield.
Ordering information: Each column is supplied with a 20 µm polyethylene bed support and ¹/₄"-28 fittings.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty