Would you like to log in again?
Chromatography Glass Columns with Removable Fritted Disc, Kimble Chase
Supplier: DWK Life Sciences
422230-2525 422230-2512 422250-2512 422230-5012 422250-2525
KT422230-2525EA 503.23 USD InStock
KT422230-2525 KT422230-2512 KT422250-2525 KT422250-2512 KT422230-5012
Chromatography Glass Columns with Removable Fritted Disc, Kimble Chase
Chromatography Columns
This column with a removable 40 to 60 µm porosity fritted disc features minimal dead space to eliminate the possible remixing of eluted components.
Complete set-up consists of a column barrel, a stopcock adapter with a size 2 PTFE plug, two ethylene propylene (EP) O-rings, a fritted disc and a pinch clamp.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty