Would you like to log in again?
KIMBLE® KONTES® Allihn Condensers, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMBLE®
431000-2930 431000-2920 431000-2440 431000-2425 431000-2430 431000-2420
KT431000-2930EA 322.01 USD InStock
KT431000-2930 KT431000-2425 10000-502 KT431000-2420 KT431000-2440 KT431000-2430
KIMBLE® KONTES® Allihn Condensers, DWK Life Sciences
Condensers
This borosilicate glass Allihn condenser are used in many refluxing operations.
  • Standard taper drip joint at bottom and a standard taper outer joint at top
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty