Would you like to log in again?
KIMBLE® KONTES® Friedrich Condensers, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMBLE®
437000-2942 456250-0021 437000-2440 456250-0023 456250-0022
10000-504EA 368.6 USD InStock
10000-504 KT456250-0021 KT437000-2440 KT456250-0023 KT456250-0022
KIMBLE® KONTES® Friedrich Condensers, DWK Life Sciences
Condensers
These Friedrich condenser specially designed to provide a long vapor path, good heat transfer and anti-flooding
characteristics, used primarily in reflux mode and Soxhlet extractions.
  • Water inlet and outlet located above ring-sealed inner condensing tube
  • Molded inner spiral provides surface area for the condensation of the product
Condenser KT437000-2440, KT437000-2942 inclined standard taper outer joint on the side and standard taper inner drip joint at the bottom.
Condenser KT456250-0022, KT456250-0021, KT456250-0023 features standard taper inner drip joint and side outlet for drying tube and more.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty