Would you like to log in again?
KONTES® Graham Condensers, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KONTES®
439000-2420 439000-2430 439000-2440 439000-2425
KT439000-2430EA 365.98 USD InStock
KT439000-2430 KT439000-2440 KT439000-2420 KT439000-2425
KONTES® Graham Condensers, Kimble Chase
Condensers
Vapor travels through a coil extending through the length of the condenser and is surrounded by a cooling jacket
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Standard Taper outer joint at top and standard taper inner drip joint at the bottom.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty