Would you like to log in again?
KONTES® Allihn Condensers, Full Length, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KONTES®
456000-0025 456000-0023 431500-2430 456000-0021 456000-0024 456000-0022
KT456000-0023EA 430.68 USD InStock
KT456000-0023 KT456000-0024 KT456000-0021 KT456000-0022 KT456000-0025 KT431500-2430
KONTES® Allihn Condensers, Full Length, Kimble Chase
Condensers
Full-length reflux condensers have a straight outlet tube at the top and a water-cooled [ST] inner drip joint at bottom
Designed for use in multiple refluxing operations including Soxhlet extractions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty