Would you like to log in again?
KONTES® Distillation Column, Vigreux, Kimble Chase
Supplier: Kimble Chase
KONTES®
503500-0223 503500-0222 503500-0221 286700-0000 286710-0300
KT286700-0000EA 168.27 USD InStock
KT286700-0000 KT286710-0300 KT503500-0221 KT503500-0222 KT503500-0223
KONTES® Distillation Column, Vigreux, Kimble Chase
Distillation Components Distillation Columns
Vigreux distilling column with indentations over the entire length for improved vapor-liquid contact.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty