Would you like to log in again?
KIMBLE® Variable Reflux Distillation Head with PTFE Plugs, Small, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
518530-2440
KT518530-2440EA 1571.31 USD InStock
KT518530-2440
KIMBLE® Variable Reflux Distillation Head with PTFE Plugs, Small, DWK Life Sciences
Distillation Components Distillation Heads
Rotation of cold finger condenser allows collection of distillate or return to flask.
  • Stopcocks allow removal of the product or a receiver change without interrupting the distillation
  • Lower receiver joint has a drip tip
  • Standard Taper 10/30 outer side joint accepts a 75 mm immersion thermometer
  • Stopcock plugs are PTFE
  • Manufactured from 33 expansion, low extractable borosilicate glass
Top and bottom plugs are KT821001-0004 and the side is KT824001-0004.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty