Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMBLE® Drying Tubes, DWK Life Sciences

Supplier: DWK Life Sciences
KIMBLE®
562500-2440
KT562500-2440EA 125.69 USD InStock
KT562500-2440
KIMBLE® Drying Tubes, DWK Life Sciences
Tubes Drying Tubes
U-shaped drying tube.

  • Inner joint at the bottom with a bulb desiccant chamber
  • Manufactured from 33 expansion, low extractable borosilicate glass

Joint length: full, overall height: 185 mm, overall width: 80 mm

Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Delivery information: Supplied with a bent inlet tube.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty