Would you like to log in again?
KIMBLE® Soxhlet Extraction Apparatus with Enlarged Vapor Tube, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
585050-0023 585050-0022
KT585050-0023CS 1091.33 USD InStock
KT585050-0023 KT585050-0022
KIMBLE® Soxhlet Extraction Apparatus with Enlarged Vapor Tube, DWK Life Sciences
Extractors Soxhlet and Soxhlet-Type Extractors
Soxhlet-type extraction apparatus with a bulb-type Allihn condenser, used for the continuous extraction of solids with a suitable solvent.
  • Standard Taper joints connect the condenser to the extractor and the extractor to the flask
  • Uses paper, alundum or all-glass extraction thimbles, not supplied
  • Manufactured from 33 expansion, low extractable borosilicate glass
Improved design with a protected siphon tube, an enlarged vapor tube and sloping seals to prevent the trapping of liquid.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty