Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
U-Shaped Spargers, Analytical Purge and Trap, Kimble Chase
Supplier: Kimble Chase
KIMBLE®
591132-2605 591132-2625
KT591132-2625EA 246.45 USD InStock
KT591132-2625 KT591132-2605
U-Shaped Spargers, Analytical Purge and Trap, Kimble Chase
Stirrers Stirring Tools
These U-shaped fritted spargers are designed for use with O-I analytical purge and trap sample concentrators.
  • Recommended for potable water and wastewater samples
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty