Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
KONTES® Needle Spargers, Purge and Trap, Kimble Chase
Supplier: DWK Life Sciences
KONTES®
591150-0005
KT591150-0005EA 43.89 USD InStock
KT591150-0005
KONTES® Needle Spargers, Purge and Trap, Kimble Chase
Stirrers Stirring Tools
Designed for use with Tekmar® 2000 and 3000 purge and trap concentrators and 2016 and 2032 autosamplers for analysis of slurries, soils, viscous liquids, and foaming liquid samples.
These spargers are also used for headspace sweep analysis of volatile compounds in oils.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty