Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Supplier: DWK Life Sciences
601500-0124 601500-0529 601500-0624 601500-1024 601500-0824 601500-2124 601500-0224 601500-0524 601500-0545 601500-0424 601500-0845 601500-0829 601500-0324 601500-0429 601500-0929
KT601500-0424EA 116.94 USD InStock
KT601500-0424 KT601500-0524 KT601500-0624 KT601500-0845 KT601500-0124 KT601500-0224 KT601500-0324 46200-630 KT601500-1024 KT601500-2124 KT601500-0829 KT601500-0929 KT601500-0824 KT601500-0429 KT601500-0529
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Flasks Flat Bottom Flasks
Single neck flat bottom flask with a Standard Taper outer joint.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty