Would you like to log in again?
KIMBLE® Distilling Flasks, Round Bottom, 2-Neck, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
605000-0624 605000-1424 605000-1824 605000-1024 605000-0224
KT605000-0624EA 176.38 USD InStock
KT605000-0624 KT605000-1824 KT605000-0224 KT605000-1024 KT605000-1424
KIMBLE® Distilling Flasks, Round Bottom, 2-Neck, DWK Life Sciences
Flasks Distillation Flasks
Distilling round bottom flask with two vertical necks.
  • Heavy wall
  • Manufactured from 33 expansion, low extractable borosilicate glass
With standard taper outer joints on both vertical necks.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty