Would you like to log in again?
KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
617000-0629 296500-0005 617000-0645 617000-0124 296500-0025 296500-0125 296500-0050 296500-0015 617000-0834 296500-0100 617000-0845 296510-0125 617000-0745 617000-1045 617000-1024 617000-0824 617000-0724 617000-0429 296510-0025 296510-0050 617000-1124 296510-0100 296510-0250 617000-1145 296500-0010
KT296500-0025EA 63.34 USD InStock
KT296500-0025 KT296500-0125 KT296500-0005 KT617000-0824 KT617000-0429 KT617000-0745 KT617000-0845 KT617000-0724 KT617000-0629 KT617000-0124 KT296500-0010 KT617000-0645 KT296500-0050 10000-516 10000-514 10000-512 KT296500-0015 10000-510 KT296510-0125 KT617000-0834 KT617000-1124 KT617000-1045 KT296500-0100 KT617000-1024 KT617000-1145
KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
Flasks Erlenmeyer Flasks
Single neck flask with a standard taper outer joint.
  • White ceramic enamel marking spot
  • Manufactured from 33 expansion, low extractable borosilicate glass
Tops are reinforced and tooled with a rounded finish, containing more glass to give them maximum mechanical strength. Body is thick-walled, with a long tapered outside contour to minimize chipping when struck or rubbed together.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty