Would you like to log in again?
KIMBLE® Atmospheric Environmental Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
653200-2000 653210-2000
KT653210-2000CS 754.99 USD InStock
KT653210-2000 KT653200-2000
KIMBLE® Atmospheric Environmental Bottles, DWK Life Sciences
Bottles Gas Washing Bottles
The design of the atmospheric environmental bottle is recommended by the Atmospheric Environment Service (AES).
  • KimCote® plastic safety coating
  • Borosilicate glass high vacuum plug valve stems instead of PTFE stems, eliminating the chance of outgassing
  • FKM valve stem o-rings that are easily changed
  • Inlet stems ground to ¹/₂" Ext.Ø
  • Manufactured from 33 expansion, low extractable borosilicate glass
Available in either a single or double valve configuration.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty