Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMBLE® Gas Washing Bottles, with Fritted Cylinder, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

KIMBLE®
657250-1223 657250-1225 657250-2525 657250-5025 657250-2523 657250-5023
KT657250-1223EA 436.24 USD InStock
KT657250-1223 KT657250-2523 KT657250-1225 KT657250-2525 KT657250-5023 KT657250-5025
KIMBLE® Gas Washing Bottles, with Fritted Cylinder, DWK Life Sciences
Bottles Gas Washing Bottles
Tall form bottles feature a coarse or extra coarse porosity fritted cylinder to maximize dispersion, 29/42 standard taper joints.

  • 2 hose connectors
  • Manufactured from 33 expansion, low extractable borosilicate glass

Supplied complete with hooks and two 5.1 cm (2") springs.

Unit contains a fritted disc for more uniform gas distribution throughout the absorbing material.

Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty