Would you like to log in again?
KIMBLE® Gas Washing Bottles, with Fritted Cylinder, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
657250-1223 657250-1225 657250-2525 657250-5025 657250-2523 657250-5023
KT657250-1223EA 444.96 USD InStock
KT657250-1223 KT657250-2523 KT657250-1225 KT657250-2525 KT657250-5023 KT657250-5025
KIMBLE® Gas Washing Bottles, with Fritted Cylinder, DWK Life Sciences
Bottles Gas Washing Bottles
Tall form bottles feature a coarse or extra coarse porosity fritted cylinder to maximize dispersion, 29/42 standard taper joints.
  • 2 hose connectors
  • Manufactured from 33 expansion, low extractable borosilicate glass
Supplied complete with hooks and two 5.1 cm (2") springs.
Unit contains a fritted disc for more uniform gas distribution throughout the absorbing material.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty